Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-(Oxiran-2-ylmethoxy)benzaldehyde
3-(Oxiran-2-ylmethoxy)benzaldehyde
3-(Oxiran-2-ylmethoxy)benzaldehyde
3-(Oxiran-2-ylmethoxy)benzaldehyde
Leverandør:
Apollo Scientific
Sertifikat
Expand image
3-(Oxiran-2-ylmethoxy)benzaldehyde
Leverandør:
Apollo Scientific
Om dette produktet
CAS-nummer:
22590-64-3
:
Formel:
C₁₀H₁₀O₃
MDL-nummer:
MFCD00203974
:
178.19 g/mol
:
GHS07
Lagringstemperatur:
Ambient