Order Entry
Norway
ContactUsLinkComponent
Cyclopentanecarboxylic acid
Cyclopentanecarboxylic acid
Catalog # APOSOR18358-100G
Supplier:  Apollo Scientific
CAS Number:  
Cyclopentanecarboxylic acid
Catalog # APOSOR18358-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :114.14 g/mol
Boiling Pt:213…216 °C
Melting Pt:44319 °C
Density:1.053 g/cm³ (25 °C)
Flash Pt:94 °C
Storage Temperature:Ambient
EINECS:222-269-5
 :GHS07
CAS Number:3400-45-1
Formula:C₆H₁₀O₂
MDL Number:MFCD00001371