Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(R)-(-)-1-Boc-3-hydroxypyrrolidine
(R)-(-)-1-Boc-3-hydroxypyrrolidine
(R)-(-)-1-Boc-3-hydroxypyrrolidine
:
Apollo Scientific
(R)-(-)-1-Boc-3-hydroxypyrrolidine
:
Apollo Scientific
:
109431-87-0
:
:
C₉H₁₇NO₃
:
MFCD01317838
:
187.24 g/mol
:
62…65 °C
:
3077
:
9+EHS,III
:
GHS07
:
GHS09
:
Ambient