
tert-Butyl-4-(chloromethyl)tetrahydro-1(2H)-pyridinecarboxylate
Catalog # APOSOR15054-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:233.74 g/mol
Melting Pt:49…51 °C
:GHS07
Storage Temperature:Ambient
CAS Number:479057-79-9
Formula:C₁₁H₂₀ClNO₂
MDL Number:MFCD08689676