Order Entry
Norway
ContactUsLinkComponent
4-Chloro-m-toluidine 98%
Irritant
Health
4-Chloro-m-toluidine 98%
Catalog # APOSOR15033-100G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Health
4-Chloro-m-toluidine 98%
Catalog # APOSOR15033-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :141.6 g/mol
Melting Pt:82…85 °C
Storage Temperature:Ambient
UN:2239
ADR:6.1,III
 :GHS07
 :GHS08
CAS Number:7149-75-9
Formula:C₇H₈ClN
MDL Number:MFCD00066332