Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Logg inn
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Hjem
2,4,6-Tribromoanisole
2,4,6-Tribromoanisole
2,4,6-Tribromoanisole
2,4,6-Tribromoanisole
Leverandør:
Apollo Scientific
Sertifikat
Expand image
2,4,6-Tribromoanisole
Leverandør:
Apollo Scientific
Om dette produktet
CAS-nummer:
607-99-8
:
Formel:
C₇H₅Br₃O
MDL-nummer:
MFCD00192510
:
344.83 g/mol
Kokende Pt:
297…299 °C
Molten Pt:
84…88 °C
:
GHS07
Lagringstemperatur:
Ambient