Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Fmoc-Thi-OH, Fmoc-3-L-Ala(3-Thienyl)-OH
Fmoc-Thi-OH, Fmoc-3-L-Ala(3-Thienyl)-OH 97%
Fmoc-Thi-OH, Fmoc-3-L-Ala(3-Thienyl)-OH 97%
Fmoc-Thi-OH, Fmoc-3-L-Ala(3-Thienyl)-OH 97%
Supplier:
Apollo Scientific
Certificates
Expand image
Fmoc-Thi-OH, Fmoc-3-L-Ala(3-Thienyl)-OH 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
186320-06-9
:
Formula:
C₂₂H₁₉NO₄S
MDL Number:
MFCD00151916
:
393.4 g/mol
Storage Temperature:
Refrigerator