Order Entry
Norway
Orders LinkContactUsLinkComponent
 

Boc-D-Tic-OH

Bottle, 250 mg
Catalog # APOSOR14588-250MG
Irriterende
Supplier:   Apollo Scientific
Boc-D-Tic-OH
Catalog # APOSOR14588-250MG
Supplier:  Apollo Scientific
CAS Number:  115962-35-1
Boc-D-Tic-OH

Bottle, 250 mg

CAS Number:  115962-35-1
Price
kr 270,00
Log in for å se tilgjengelighet og deres pris
1 * 250 mg
Product Details & Documents
Synonymer:  Boc-D-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid
  • CAS Number:115962-35-1
  • Formel:C₁₅H₁₉NO₄
  • MDL-nummer:MFCD00143818
  • Molecular Weight:277.32 g/mol
  • Lagringstemperatur:Ambient
Spesifikasjoner

Spesifikasjoner

  • Pack type:
    Bottle
  • Pk:
    250 mg

Spesifikasjoner

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.