kr 0,00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Norway
ContactUsLinkComponent
Fmoc-4-chloro-L-phenylalanine
Fmoc-4-chloro-L-phenylalanine 97%
Fmoc-4-chloro-L-phenylalanine 97%
APOSOR14550-5G
:
Apollo Scientific
:
Fmoc-4-chloro-L-phenylalanine 97%
APOSOR14550-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
421.877 g/mol
:
Ambient
:
175453-08-4
:
C₂₄H₂₀ClNO₄
:
MFCD00080281