Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
N-Boc-L-prolinol
N-Boc-L-prolinol
N-Boc-L-prolinol
Irriterende
N-Boc-L-prolinol
Leverandør:
Apollo Scientific
Sertifikat
Utvid bildet
Irriterende
N-Boc-L-prolinol
Leverandør:
Apollo Scientific
Om dette produktet
CAS-nummer:
69610-40-8
:
Formel:
C₁₀H₁₉NO₃
MDL-nummer:
MFCD00066232
:
201.27 g/mol
Kokende Pt:
150 °C/4mmHg
Molten Pt:
62…64 °C
:
GHS07
Lagringstemperatur:
Ambient