
[3-(4-Methoxyphenyl)-5-isoxazolyl]methanol
Catalog # APOSOR14216-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
Melting Pt:97…98 °C
:GHS07
Storage Temperature:Ambient
CAS Number:206055-86-9
Formula:C₁₁H₁₁NO₃
MDL Number:MFCD04124287