Order Entry
Norway
ContactUsLinkComponent
3,4,5-Trimethoxybenzoic acid
Irritant
3,4,5-Trimethoxybenzoic acid
Catalog # APOSOR13042-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
3,4,5-Trimethoxybenzoic acid
Catalog # APOSOR13042-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :212.2 g/mol
Boiling Pt:225…227 °C/10mmHg
Melting Pt:168…171 °C
EINECS:204-248-2
 :GHS07
Storage Temperature:Ambient
CAS Number:118-41-2
Formula:C₁₀H₁₂O₅
MDL Number:MFCD00002501