
2-Bromo-1-(2-pyridinyl)-1-ethanone hydrobromide 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:17570-98-8
:
Formula:C₇H₇Br₂NO
MDL Number:MFCD02181196
:280.96 g/mol
Melting Pt:205…207 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together