
[3-Phenyl-5-isoxazolyl]methanol 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:90924-12-2
:
Formula:C₁₀H₉NO₂
MDL Number:MFCD01928812
:175.1840 g/mol
Melting Pt:45…45 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS07
Frequently Bought Together