kr 0,00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Norway
ContactUsLinkComponent
5-Amino-2-chlorophenol
5-Amino-2-chlorophenol
5-Amino-2-chlorophenol
:
Apollo Scientific
5-Amino-2-chlorophenol
:
Apollo Scientific
:
6358-06-1
:
:
C₆H₆ClNO
:
MFCD01707858
:
143.57 g/mol
:
160 °C
:
GHS07
:
Kjøleskap