kr 0,00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Norway
ContactUsLinkComponent
1,3-Dibromo-5-nitrobenzene
1,3-Dibromo-5-nitrobenzene 97%
1,3-Dibromo-5-nitrobenzene 97%
1,3-Dibromo-5-nitrobenzene 97%
Supplier:
Apollo Scientific
Certificates
Expand image
1,3-Dibromo-5-nitrobenzene 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
6311-60-0
:
Formula:
C₆H₃Br₂NO₂
MDL Number:
MFCD00092529
:
280.9 g/mol
Melting Pt:
100…104 °C
:
GHS07
Storage Temperature:
Ambient