
3-Nitrobenzenesulphonyl chloride 96%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:121-51-7
:
Formula:C₆H₄ClNO₄S
MDL Number:MFCD00007435
Melting Pt:61…62 °C
Storage Temperature:Ambient
EINECS:204-476-2
UN:3261
ADR:8,II
:221.62 g/mol
:GHS05