Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Chloro-1-methylimidazole
2-Chloro-1-methylimidazole 95%
2-Chloro-1-methylimidazole 95%
APOSOR10413-1G
:
Apollo Scientific
:
2-Chloro-1-methylimidazole 95%
APOSOR10413-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
116.55 g/mol
:
GHS07
:
Ambient
:
253453-91-7
:
C₄H₅ClN₂
:
MFCD02179531