Skip to Header
Skip to Main Content
Skip to Footer
Avansert
Avansert
SDS
Sertifikat
Produsentdata
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl propiolate
Methyl propiolate
Methyl propiolate
:
Apollo Scientific
Methyl propiolate
:
Apollo Scientific
:
922-67-8
:
:
C₄H₄O₂
:
MFCD00008572
:
103…105 °C
:
0.945 g/cm³ (25 °C)
:
10 °C
:
213-083-5
:
3272
:
3,II
:
84.07 g/mol
:
GHS02
:
GHS07
:
Ambient