Order Entry
Norway
Orders LinkContactUsLinkComponent
4'-Bromoacetophenone

4'-Bromoacetophenone

Irritant
Health
Supplier:   Apollo Scientific
4'-Bromoacetophenone
Supplier:  Apollo Scientific
Certificates
4'-Bromoacetophenone

2 Options Available Below

Product Details & Documents
Synonyms:  p-Bromoacetophenone, 1-(4-Bromophenyl)ethanone
  • CAS Number:99-90-1
  • Formula:C₈H₇BrO
  • MDL Number:MFCD00000105
  • Molecular Weight:199.05 g/mol
  • Boiling Pt:255…256 °C
  • Melting Pt:49…53 °C
  • Density:1.647 g/cm³ (25 °C)
  • Flash Pt:>110 °C
  • Storage Temperature:Ambient
  • EINECS:202-799-3
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - kr 270,00
    kr 270,00
    1 * 100 g
       kr 270,00

  • Bottle
    Log in for availability
    Log in for availability
    1 * 250 g
    1 * 250 g - kr 360,00
    kr 360,00
    1 * 250 g
       kr 360,00

    Specifications:
    New Product
    New Product