2-Amino-3,5-dibromopyrazine 99%
Catalog # APOSOR01487-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:252.90 g/mol
Melting Pt:114…117 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS06
CAS Number:24241-18-7
Formula:C₄H₃Br₂N₃
MDL Number:MFCD00673150