Order Entry
Norway
ContactUsLinkComponent
Sodium tungstate dihydrate 98-101% ACS
Sodium tungstate dihydrate 98-101% ACS
  APOSIN3275-100G
 :  
Sodium tungstate dihydrate 98-101% ACS
  APOSIN3275-100G
 :  
  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401
 :
 :329.85 g/mol
 :698 °C
 :3.23…3.25 g/cm³ (25 °C)
 :236-743-4
 :GHS07
 :Ambient
 :10213-10-2
 :Na₂WO₄·2H₂O
 :MFCD00149190