Order Entry
Norway
ContactUsLinkComponent
Rubidium chloride 99%
Rubidium chloride 99%
Catalog # APOSIN3019-25G
Supplier:  Apollo Scientific
CAS Number:  
Rubidium chloride 99%
Catalog # APOSIN3019-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :120.92 g/mol
Boiling Pt:1390 °C
Melting Pt:715 °C
Density:2.8 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:232-240-9
 :GHS07
CAS Number:7791-11-9
Formula:RbCl
MDL Number:MFCD00011187