Cefaclor
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: (6R,7R)-7-((R)-2-Amino-2-phenylacetamido)-3-chloro-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
- CAS Number:53994-73-3
- Formula:C₁₅H₁₄ClN₃O₄S
- MDL Number:MFCD00151471
- Molecular Weight:385.8 g/mol
- Storage Temperature:Ambient
- EINECS:258-909-5
3 Options Available Below
Product Details & Documents
Synonyms: (6R,7R)-7-((R)-2-Amino-2-phenylacetamido)-3-chloro-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
- CAS Number:53994-73-3
- Formula:C₁₅H₁₄ClN₃O₄S
- MDL Number:MFCD00151471
- Molecular Weight:385.8 g/mol
- Storage Temperature:Ambient
- EINECS:258-909-5
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



