kr 0,00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Norway
ContactUsLinkComponent
Gö 6983
Gö 6983 ≥99%
Gö 6983 ≥99%
:
Apollo Scientific
Gö 6983 ≥99%
:
Apollo Scientific
:
133053-19-7
:
:
C₂₆H₂₆N₄O₃
:
MFCD04040031
:
442.519 g/mol
:
GHS07
:
Freezer