Djenkolic acid
Catalog # APOSBID1201-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:254.32 g/mol
EINECS:207-863-4
:GHS07
Storage Temperature:Refrigerator
CAS Number:498-59-9
Formula:C₇H₁₄N₂O₄S₂
MDL Number:MFCD00063079