Order Entry
Norway
ContactUsLinkComponent
BMPS (3-Maleimidopropionic acid N-hydroxysuccinimide ester)
Irritant
BMPS (3-Maleimidopropionic acid N-hydroxysuccinimide ester)
Catalog # APOSBICL228-100MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
BMPS (3-Maleimidopropionic acid N-hydroxysuccinimide ester)
Catalog # APOSBICL228-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :266.21 g/mol
Melting Pt:168…170 °C
 :GHS07
Storage Temperature:Refrigerator
CAS Number:55750-62-4
Formula:C₁₁H₁₀N₂O₆
MDL Number:MFCD00043141