Cerulenin
Catalog # APOSBIC1007-5MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 mg
- Seq:2210
Specifications
About this item
:
:223.272 g/mol
Melting Pt:93 °C
EINECS:241-424-8
:GHS07
Storage Temperature:Freezer
CAS Number:17397-89-6
Formula:C₁₂H₁₇NO₃
MDL Number:MFCD00077686