Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Clindamycin hydrochloride
Clindamycin hydrochloride
Clindamycin hydrochloride
Clindamycin hydrochloride
Supplier:
Apollo Scientific
Certificates
Utvid bildet
Clindamycin hydrochloride
Supplier:
Apollo Scientific
Certificates
About this item
CAS Number:
21462-39-5
:
Formula:
C₁₈H₃₃ClN₂O₅S·HCl
MDL Number:
MFCD07793327
:
461.45 g/mol
:
GHS07
Storage Temperature:
Refrigerator
Related Information
Dokumentasjon
Certificates
Search by Lot Number
Dokumentasjon