Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,2'-Bicinchoninic acid disodium salt
2,2'-Bicinchoninic acid disodium salt ≥98%
2,2'-Bicinchoninic acid disodium salt ≥98%
Irritant
Health
2,2'-Bicinchoninic acid disodium salt ≥98%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
Health
2,2'-Bicinchoninic acid disodium salt ≥98%
Supplier:
Apollo Scientific
Certificates
About this item
CAS Number:
979-88-4
:
Formula:
C₂₀H₁₀N₂Na₂O₄
MDL Number:
MFCD00037500
:
388.29 g/mol (anhy)
:
GHS07
:
GHS08
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
Documentation