kr 0,00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Norway
ContactUsLinkComponent
4-Fluoro-2-formylbenzoic acid
4-Fluoro-2-formylbenzoic acid 98%
4-Fluoro-2-formylbenzoic acid 98%
Irritant
4-Fluoro-2-formylbenzoic acid 98%
Supplier:
Apollo Scientific
Certificates
Utvid bildet
Irritant
4-Fluoro-2-formylbenzoic acid 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
1186047-15-3
:
Formula:
C₈H₅FO₃
MDL Number:
MFCD20658188
:
168.122 g/mol
:
GHS07
Storage Temperature:
Ambient