Order Entry
Norway
ContactUsLinkComponent
 
5-Fluoro-2-nitrobenzenesulphonyl chloride 97%
Catalog # APOSPC501559-1G
Supplier:  Apollo Scientific
CAS Number:  
5-Fluoro-2-nitrobenzenesulphonyl chloride 97%
Catalog # APOSPC501559-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
ADR:8,III
 :239.609 g/mol
Storage Temperature:Ambient
UN:1759
CAS Number:82711-97-5
Formula:C₆H₃ClFNO₄S
MDL Number:MFCD11858121