Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Amino-8-fluoroquinoline dihydrochloride
3-Amino-8-fluoroquinoline dihydrochloride
3-Amino-8-fluoroquinoline dihydrochloride
3-Amino-8-fluoroquinoline dihydrochloride
Supplier:
Apollo Scientific
Certificates
Expand image
3-Amino-8-fluoroquinoline dihydrochloride
Supplier:
Apollo Scientific
About this item
CAS Number:
1266226-18-9
:
Formula:
C₉H₇FN₂·2ClH
MDL Number:
MFCD18375334
:
235.086 g/mol
:
GHS07
Storage Temperature:
Ambient