Order Entry
Norway
ContactUsLinkComponent
2-Bromo-5-fluoro-4-methylbenzaldehyde 98%
2-Bromo-5-fluoro-4-methylbenzaldehyde 98%
Catalog # APOSPC48739-5G
Supplier:  Apollo Scientific
CAS Number:  
2-Bromo-5-fluoro-4-methylbenzaldehyde 98%
Catalog # APOSPC48739-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :217.035 g/mol
Storage Temperature:Ambient
CAS Number:916792-21-7
Formula:C₈H₆BrFO
MDL Number:MFCD07782047