Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Bromo-2-fluoro-3-methylbenzaldehyde
5-Bromo-2-fluoro-3-methylbenzaldehyde 97%
5-Bromo-2-fluoro-3-methylbenzaldehyde 97%
5-Bromo-2-fluoro-3-methylbenzaldehyde 97%
Supplier:
Apollo Scientific
Certificates
Expand image
5-Bromo-2-fluoro-3-methylbenzaldehyde 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
903875-64-9
:
Formula:
C₈H₆BrFO
MDL Number:
MFCD11110261
:
217.035 g/mol
:
GHS07
Storage Temperature:
Refrigerator