Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Sodium 2,3,4,5,6-pentafluorobenzoate
Sodium 2,3,4,5,6-pentafluorobenzoate
Sodium 2,3,4,5,6-pentafluorobenzoate
Sodium 2,3,4,5,6-pentafluorobenzoate
Supplier:
Apollo Scientific
Certificates
Expand image
Sodium 2,3,4,5,6-pentafluorobenzoate
Supplier:
Apollo Scientific
About this item
CAS Number:
4830-57-3
:
Formula:
C₇F₅O₂·Na
MDL Number:
MFCD01652013
:
234.055 g/mol
:
GHS07
Storage Temperature:
Ambient