Gå til Overskrift
Gå til Hovedinnhold
Gå til Bunntekst
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(S)-1-(3,5-Difluorophenyl)ethanol
(S)-1-(3,5-Difluorophenyl)ethanol
(S)-1-(3,5-Difluorophenyl)ethanol
Irritant
(S)-1-(3,5-Difluorophenyl)ethanol
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
(S)-1-(3,5-Difluorophenyl)ethanol
Supplier:
Apollo Scientific
About this item
CAS Number:
692732-03-9
:
Formula:
C₈H₈F₂O
MDL Number:
MFCD18072180
:
158.145 g/mol
Flash Pt:
68 °C
:
GHS07
Storage Temperature:
Ambient