Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
N,O-Bis(2,2,2-trifluoroethyl)-4-aminophenol
N,O-Bis(2,2,2-trifluoroethyl)-4-aminophenol
N,O-Bis(2,2,2-trifluoroethyl)-4-aminophenol
N,O-Bis(2,2,2-trifluoroethyl)-4-aminophenol
Supplier:
Apollo Scientific
Certificates
Expand image
N,O-Bis(2,2,2-trifluoroethyl)-4-aminophenol
Supplier:
Apollo Scientific
About this item
CAS Number:
1309602-67-2
:
Formula:
C₁₀H₉F₆NO
MDL Number:
MFCD18074427
:
273.175 g/mol
Storage Temperature:
Ambient