Order Entry
Norway
ContactUsLinkComponent
Ethyl 1-(4-(aminosulphonyl)phenyl)-3-(trifluoromEthyl)-5-p-tolyl-1H-pyrazole-4-carboxylate
Ethyl 1-(4-(aminosulphonyl)phenyl)-3-(trifluoromEthyl)-5-p-tolyl-1H-pyrazole-4-carboxylate
Catalog # APOSPC430599-500MG
Supplier:  Apollo Scientific
CAS Number:  
Ethyl 1-(4-(aminosulphonyl)phenyl)-3-(trifluoromEthyl)-5-p-tolyl-1H-pyrazole-4-carboxylate
Catalog # APOSPC430599-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pakketype:
    Bottle
  • Fp:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :453.435 g/mol
Storage Temperature:Romtemperatur
CAS Number:1206970-24-2
Formula:C₂₀H₁₈F₃N₃O₄S
MDL Number:MFCD14585471