Order Entry
Norway
ContactUsLinkComponent
2-[2-(6-Fluoroimidazo[1,2-a]pyridin-2-yl)ethyl]-1H-isoindole-1,3(2H)-dione hydrobromide
2-[2-(6-Fluoroimidazo[1,2-a]pyridin-2-yl)ethyl]-1H-isoindole-1,3(2H)-dione hydrobromide
Catalog # APOSPC300769-1G
Supplier:  Apollo Scientific
CAS Number:  
2-[2-(6-Fluoroimidazo[1,2-a]pyridin-2-yl)ethyl]-1H-isoindole-1,3(2H)-dione hydrobromide
Catalog # APOSPC300769-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :390.206 g/mol
Storage Temperature:Ambient
CAS Number:1858250-77-7
Formula:C₁₇H₁₂FN₃O₂·BrH
MDL Number:MFCD27995622