Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-Chloro-3-fluoro-5-nitrobenzene
1-Chloro-3-fluoro-5-nitrobenzene 99%
1-Chloro-3-fluoro-5-nitrobenzene 99%
Irritant
1-Chloro-3-fluoro-5-nitrobenzene 99%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
1-Chloro-3-fluoro-5-nitrobenzene 99%
Supplier:
Apollo Scientific
About this item
CAS Number:
4815-64-9
:
Formula:
C₆H₃ClFNO₂
MDL Number:
MFCD11973918
:
175.545 g/mol
:
GHS07
Storage Temperature:
Ambient