

4-Methyl-5,5,5-trifluoropent-1-en-4-ol
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:73893-33-1
:
Formula:C₆H₉F₃O
MDL Number:MFCD07784257
:154.13 g/mol
Boiling Pt:108…110 °C
UN:1993
ADR:3,III
:GHS02
:GHS07
Storage Temperature:Ambient