Order Entry
Norway
ContactUsLinkComponent
5-(4-Methoxyphenyl)-3H-1,3,4-oxadiazol-2-one 98%
5-(4-Methoxyphenyl)-3H-1,3,4-oxadiazol-2-one 98%
Catalog # APOSOR954532-1G
Supplier:  Apollo Scientific
CAS Number:  
5-(4-Methoxyphenyl)-3H-1,3,4-oxadiazol-2-one 98%
Catalog # APOSOR954532-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :192.171 g/mol
Storage Temperature:Ambient
CAS Number:41125-75-1
Formula:C₉H₈N₂O₃
MDL Number:MFCD08669523