Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-Methyl-1H-indol-6-amine
1-Methyl-1H-indol-6-amine 96%
1-Methyl-1H-indol-6-amine 96%
APOSOR953134-1G
:
Apollo Scientific
:
1-Methyl-1H-indol-6-amine 96%
APOSOR953134-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
146.189 g/mol
:
Ambient
:
135855-62-8
:
C₉H₁₀N₂
:
MFCD10000822