Order Entry
Norway
ContactUsLinkComponent
tert-Butyl-4-methoxycarbanilate 95%
tert-Butyl-4-methoxycarbanilate 95%
Catalog # APOSOR946958-5G
Supplier:  Apollo Scientific
CAS Number:  
tert-Butyl-4-methoxycarbanilate 95%
Catalog # APOSOR946958-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :223.268 g/mol
Storage Temperature:Ambient
CAS Number:18437-68-8
Formula:C₁₂H₁₇NO₃
MDL Number:MFCD00816766