kr 0,00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Norway
ContactUsLinkComponent
5-(1-Piperidinyl)-3-pentyn-1-ol
5-(1-Piperidinyl)-3-pentyn-1-ol 95%
5-(1-Piperidinyl)-3-pentyn-1-ol 95%
5-(1-Piperidinyl)-3-pentyn-1-ol 95%
Supplier:
Apollo Scientific
Certificates
Expand image
5-(1-Piperidinyl)-3-pentyn-1-ol 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
104580-60-1
:
Formula:
C₁₀H₁₇NO
MDL Number:
MFCD02012810
:
167.248 g/mol
Storage Temperature:
Ambient