Order Entry
Norway
ContactUsLinkComponent
4,6-Dihydroxy-5-methyl-1,3-diformyl benzene 95%
4,6-Dihydroxy-5-methyl-1,3-diformyl benzene 95%
Catalog # APOSOR933368-1G
Supplier:  Apollo Scientific
CAS Number:  
4,6-Dihydroxy-5-methyl-1,3-diformyl benzene 95%
Catalog # APOSOR933368-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :180.157 g/mol
Storage Temperature:Ambient
CAS Number:22304-67-2
Formula:C₉H₈O₄
MDL Number:MFCD11505923