Skip to Header
Skip to Main Content
Skip to Footer
Avansert
Avansert
SDS
Sertifikat
Produsentdata
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-(4-Amino-2-methoxy-phenyl)-ethanone
1-(4-Amino-2-methoxy-phenyl)-ethanone 95%
1-(4-Amino-2-methoxy-phenyl)-ethanone 95%
1-(4-Amino-2-methoxy-phenyl)-ethanone 95%
APOSOR918133-1G
:
Apollo Scientific
:
1-(4-Amino-2-methoxy-phenyl)-ethanone 95%
APOSOR918133-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
165.189 g/mol
:
Ambient
:
60207-18-3
:
C₉H₁₁NO₂
:
MFCD11848247