Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-Methylpyridine-2,3-dicarboxylic acid
6-Methylpyridine-2,3-dicarboxylic acid 96%
6-Methylpyridine-2,3-dicarboxylic acid 96%
Irritant
6-Methylpyridine-2,3-dicarboxylic acid 96%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
6-Methylpyridine-2,3-dicarboxylic acid 96%
Supplier:
Apollo Scientific
About this item
CAS Number:
53636-70-7
:
Formula:
C₈H₇NO₄
MDL Number:
MFCD00075289
:
181.145 g/mol
Melting Pt:
170 °C (dec.)
:
GHS07
Storage Temperature:
Ambient