

Methyl (2E)-4-methylpent-2-enoate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:20515-15-5
:
Formula:C₇H₁₂O₂
MDL Number:MFCD00137612
:128.169 g/mol
Boiling Pt:74…75 °C (46 mm Hg)
Flash Pt:47 °C
Storage Temperature:Ambient
UN:1993
ADR:3,III
:GHS02
:GHS07